Learn More
Cayman Chemical Asarone 10mg
SDP
Catalog No. 502951972
Encompass_Preferred
Ths mxture contans the prmary COX products produced by most mammalan tssues Contents Prostaglandn D2 Prostaglandn E2 6keto Prostaglandn F1a Prostaglandn F2a and Thromboxane B2 (100 Ug each)
Catalog No. 50-295-1972 Supplier Cayman Chemical Supplier No. 11682 10MG
Ths mxture contans the prmary COX products produced by most mammalan tssues Contents Prostaglandn D2 Prostaglandn E2 6keto Prostaglandn F1a Prostaglandn F2a and Thromboxane B2 (100 Ug each)