Learn More
Medchemexpress LLC HY-N6826 1mg Medchemexpress, Asatone CAS:38451-63-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN68261MG
Encompass_Preferred
Medchemexpress, HY-N6826 1mg Asatone CAS:38451-63-7 Asatone is an active component isolated from Radix et Rhizoma Asari, with anti-inflammatory effect via activation of NF-κB and donwn regulation of p-MAPK (ERK, JNK and p38) pathways [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3972