Learn More
Aviva Systems Biology Corp ASB11 Antibody - C-terminal region (ARP51819_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51819_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ASB11. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human ASB11. Alternate names: DKFZp779E2460, MGC119168, MGC119169ApplicationWB. Species reactivity: Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-60624