Learn More
Aviva Systems Biology Corp ASB12 Antibody - C-terminal region (ARP52534_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52534_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ASB12. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human ASB12. Alternate names: FLJ39577ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60765