Learn More
Aviva Systems Biology Corp ASB3 Antibody - middle region (ARP57739_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57739_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ASB3. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human ASB3. Alternate names: ASB-3, FLJ10123, FLJ10421, MGC12531, MGC132002, MGC996ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Rat
Catalog No. 50-286-369