Learn More
Aviva Systems Biology Corp ASB4 Antibody - middle region (ARP62054_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62054_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ASB4. It was validated on Western Blot. . Alternate names: ASB-4, MGC142039, MGC142041ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast
Catalog No. 50-287-553