Learn More
Aviva Systems Biology Corp ASB5 Antibody - C-terminal region (ARP50276_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50276_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ASB5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human ASB5. Alternate names: ASB-5ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-60306