Learn More
Aviva Systems Biology Corp ASB8 Antibody - N-terminal region (ARP52408_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52408_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ASB8. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ASB8. Alternate names: FLJ21255, MGC5540, PP14212ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60729