Learn More
Brady™ Asbestos Warning Pipe Markers, Legend: CAUTION ASBESTOS HAZARD…..TRAINING AND EQUIPMENT
Shop All Brady Corporation ProductsDescription
- Includes industrial grade adhesive and peel-to-release coated paper backing
- Identifiy asbestos or asbestos free pipes
Specifications
Specifications
| Description | Style IHV |
| Adhesive Type | Acrylic, Permanent, Cold-temperature |
| Color | Black, Yellow |
| Format | Die-cut |
| Material | Pressure-sensitive Vinyl (B-946) |
| Display Text | CAUTION ASBESTOS HAZARD DO NOT DISTRUB WITHOUT PROPER TRAINING AND EQUIPMENT |
| Size (English) | 24 x 4 in. (W x H) |
| Size (Metric) | 60.9 x 10.16 cm (W x H) |
| Thickness (English) | 0.004 in. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.