Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Asbestos Warning Pipe Markers, Legend: CAUTION ASBESTOS HAZARD…..TRAINING AND EQUIPMENT

Catalog No. 19112021 Shop All Brady Corporation Products
Change view
Click to view available options
Description:
Style I
Style IHV
Size (English):
14 x 2.25 in. (W x H)
24 x 4 in. (W x H)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description Size (English)
19-112-021 Style I 14 x 2.25 in. (W x H)
19-112-020 Style IHV 24 x 4 in. (W x H)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-112-021 Supplier Brady™ Supplier No. 97628

To identifiy asbestos content in pipes. Brady™ Asbestos Warning Pipe Markers identify flow direction and contents at the point of need and are made of durable vinyl suitable for indoor or outdoor environments.

  • Includes industrial grade adhesive and peel-to-release coated paper backing
  • Identifiy asbestos or asbestos free pipes

Specifications

Description Style I
Adhesive Type Acrylic, Permanent, Cold-temperature
Color Black, Yellow
Format Die-cut
Material Pressure-sensitive Vinyl (B-946)
Display Text CAUTION ASBESTOS HAZARD DO NOT DISTRUB WITHOUT PROPER TRAINING AND EQUIPMENT
Size (English) 14 x 2.25 in. (W x H)
Size (Metric) 35.56 x 5.71 cm (W x H)
Thickness (English) 0.004 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.