Learn More
Enzo Life Sciences ASC polyclonal antibody (100 µg)
SDP
Supplier:  Enzo Life Sciences ADI905173100
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to aa 182-195 of human ASC. UniProt: Q9ULZ3. Species Reactivity: Human. Applications: ICC, WB. Recommended dilutions: Western Blot (1µg/ml). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Peptide affinity purified. Formulation: Liquid. In PBS containing 0.02% sodium azide.
Catalog No. 50-658-277