Learn More
AdipoGen Asc AL177 Blocking Peptide
SDP
Supplier:  AdipoGen AG37B0001C100
Encompass_Preferred
Protein. Source Synthetic. Liquid. In sterile water. Purity >95% HPLC. Blocking Peptide for anti-Asc, pAb AL177. This vial contains 100µg peptide in 100µl sterile water. The Asc Antibody AL177 Blocking Peptide can be used in conjunction with several variants of the anti-Asc pAb AL177 Prod. No. AG-25B-0006 to block protein-antibody complex formation.
Catalog No. 50-256-3065