Learn More
Novus Biologicals™ ASC/TMS1 Antibody Blocking Peptide

Specifications
Specifications
| Gene ID (Entrez) | 29108 |
| Concentration | 1 mg/mL |
| Content And Storage | Store at -80°C. Avoid freeze-thaw cycles. |
| For Use With (Application) | This peptide is useful as a blocking peptide for NBP1-78978. For further blocking peptide related protocol, click here . |
| Gene Alias | apoptosis-associated speck-like protein containing a CARD, ASC, CARD5, caspase recruitment domain protein 5, Caspase recruitment domain-containing protein 5, hASC, PYCARD, PYD and CARD domain-containing protein, Target of methylation-induced silencing 1, TMS, TMS1, TMS-1 |
| Gene Symbol | PYCARD |
| Quantity | 0.1 mg |
| Research Category | Apoptosis, Cancer, Cell Biology, Inflammation, Innate Immunity |
| Storage Buffer | Peptide dissolved in dH20. Contains no BSA. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.