Learn More
ABclonal Technology ASC/TMS1 Rabbit mAb
SDP
Catalog No. 5000007614
Encompass_Preferred
Enables identical protein binding activity and protein dimerization activity Involved in several processes including activation of cysteine-type endopeptidase activity positive regulation of cytokine production and regulation of defense response Acts upstream of or within several processes including defense response to Gram-positive bacterium positive regulation of macromolecule metabolic process and regulation of autophagy Located in cytosol extracellular region and nucleus Part of inflammasome complex Is expressed in several structures including alimentary system brain heart hemolymphoid system gland and urinary system Orthologous to human PYCARD (PYD and CARD domain containing)
Catalog No. 50-000-07614 Supplier ABclonal Technology Supplier No. A2416520UL
Enables identical protein binding activity and protein dimerization activity Involved in several processes including activation of cysteine-type endopeptidase activity positive regulation of cytokine production and regulation of defense response Acts upstream of or within several processes including defense response to Gram-positive bacterium positive regulation of macromolecule metabolic process and regulation of autophagy Located in cytosol extracellular region and nucleus Part of inflammasome complex Is expressed in several structures including alimentary system brain heart hemolymphoid system gland and urinary system Orthologous to human PYCARD (PYD and CARD domain containing)