Learn More
TARGETMOL CHEMICALS INC ASCAMYCIN 5MG
SDP
Catalog No. 502581297
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Ascamycin is a 5'-O-sulfonamide ribonucleoside antibiotic produced by Streptomyces spp. It shows selective antibacterial activity against Xanthomonas species and inhibits Xanthomonas citri Xanthomonas oryzae and phage with MIC values of 0.4 ug/mL 12.5 ug/mL and 12.5 ug/mL respectively. purity: 99%
Catalog No. 50-258-1297 Supplier TARGETMOL CHEMICALS INC Supplier No. T143295MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Ascamycin is a 5'-O-sulfonamide ribonucleoside antibiotic produced by Streptomyces spp. It shows selective antibacterial activity against Xanthomonas species and inhibits Xanthomonas citri Xanthomonas oryzae and phage with MIC values of 0.4 ug/mL 12.5 ug/mL and 12.5 ug/mL respectively. purity: 99%