Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ascaroside C3 | 1086696-26-5 | 10 MG
SDP

Catalog No. 5000418630
Encompass_Preferred

Ascr#5 (Ascaroside C3) is a highly conserved ascaroside isolated from *Caenorhabditis elegans*.

  • White to light yellow solid
  • High purity of 98.0%
  • Molecular formula C9H16O6
  • Molecular weight 220.22
  • Soluble in DMSO up to 100 mg/mL
  • In vivo dissolution protocols available

Catalog No. 50-004-18630 Supplier Medchemexpress LLC Supplier No. HYN697810MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ascr#5 (Ascaroside C3) is a highly conserved ascaroside isolated from *Caenorhabditis elegans*.

  • White to light yellow solid
  • High purity of 98.0%
  • Molecular formula C9H16O6
  • Molecular weight 220.22
  • Soluble in DMSO up to 100 mg/mL
  • In vivo dissolution protocols available

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.