Learn More
Abcam AsCas12a (Active), 100UG
Catalog No. 502703291
Encompass_Preferred
AsCas12a (Active) is a Acidaminococcus sp. BV3L6 Full Length protein, in the 1 to 1307 aa range, expressed in Escherichia coli, with >=95% purity, < 10 EU/mg endotoxin level and suitable for SDS-PAGE, Biological Activity, HPLC.
Protein/ Peptide/ Enzyme Synonyms: cpf1, HMPREF1246_0236, cas12a, CRISPR-associated endonuclease Cas12a, AsCpf1, CRISPR-associated endonuclease Cpf1
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-3291 Supplier Abcam Supplier No. AB316171100UG
AsCas12a (Active) is a Acidaminococcus sp. BV3L6 Full Length protein, in the 1 to 1307 aa range, expressed in Escherichia coli, with >=95% purity, < 10 EU/mg endotoxin level and suitable for SDS-PAGE, Biological Activity, HPLC.
Protein/ Peptide/ Enzyme Synonyms: cpf1, HMPREF1246_0236, cas12a, CRISPR-associated endonuclease Cas12a, AsCpf1, CRISPR-associated endonuclease Cpf1
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement