Learn More
Aviva Systems Biology Corp Ascc1 Antibody - C-terminal region (ARP56783_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56783_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Ascc1. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Rat. Alternate names: Asc-1, CGI-18, MGC94577, Ascc1ApplicationWB. Species reactivity: Cow, Dog, Goat, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-286-213