Learn More
New England Biolabs, Inc. AscI – 2500 units
SDP
Catalog No. 50812240
Encompass
A restriction endonuclease that recognizes the sequence GG^CGCG_CC.
Catalog No. 50-812-240 Supplier New England Biolabs, Inc. Supplier No. R0558L
A restriction endonuclease that recognizes the sequence GG^CGCG_CC.