Learn More
Medchemexpress LLC HY-104010 50mg , Asciminib CAS:1492952-76-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10401050MG
Encompass_Preferred
HY-104010 50mg Asciminib CAS: 1492952-76-7 Asciminib (ABL001) is a potent and selective allosteric BCR-ABL1 inhibitor, which inhibits Ba/F3 cells grown with an IC50 of 0.25 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9910