Learn More
Selleck Chemical LLC Asciminib (ABL001) 2mg 1492952-76-7
SDP
Supplier:  Selleck Chemical LLC S85552MG
Encompass_Preferred
Asciminib (ABL001) is a potent and selective allosteric ABL1 inhibitor with dissociation constant (Kd) of?0.5-0.8?nM and selectivity to the myristoyl pocket of ABL1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8129