Learn More
Medchemexpress LLC Asciminib hydrochlor 10mM 1mL | 2119669-71-3 | 10MM 1ML
SDP
Catalog No. 5000243500
Encompass_Preferred
Asciminib (ABL001) hydrochloride is a potent and selective allosteric BCR-ABL1 inhibitor which inhibits Ba/F3 cells grown with an IC50 of 0 25 nM[1
Catalog No. 50-002-43500 Supplier Medchemexpress LLC Supplier No. HY104010A10MM 1ML
Asciminib (ABL001) hydrochloride is a potent and selective allosteric BCR-ABL1 inhibitor which inhibits Ba/F3 cells grown with an IC50 of 0 25 nM[1