Learn More
Aviva Systems Biology Corp ASCL1 Antibody - N-terminal region (ARP32355_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32355_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ASCL1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human ASCL1. Alternate names: ASH1, HASH1, MASH1, bHLHa46ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54540