Learn More
Cell Signaling Technology ASCL1 Monoclonal Antibody, Rabbit IgG, Unconjugated, 100 μL
SDP
Catalog No. 502733024
Encompass
The ASCL1 (E6Y1B) Rabbit Monoclonal Antibody is designed to detect endogenous levels of total ASCL1 protein. This monoclonal antibody is generated by immunizing animals with a recombinant protein specific to the carboxy terminus of the human ASCL1 protein. ASCL1, also known as MASH-1, is a basic helix-loop-helix (BHLH) transcription factor essential for the differentiation of neuroendocrine cells and neural tissues. It functions as a pioneer transcription factor that accesses closed chromatin to promote the transcription of neuronal genes.
  • Validated applications include western blotting, immunoprecipitation, and immunofluorescence (immunocytochemistry).
  • Species reactivity is confirmed for human, mouse, and rat.
  • Provided in a stable formulation with glycerol for storage at −20°C.
  • Recognizes endogenous levels of the 30 kDa ASCL1 protein.
Catalog No. 50-273-3024 Supplier Cell Signaling Technology Supplier No. 55467S
The ASCL1 (E6Y1B) Rabbit Monoclonal Antibody is designed to detect endogenous levels of total ASCL1 protein. This monoclonal antibody is generated by immunizing animals with a recombinant protein specific to the carboxy terminus of the human ASCL1 protein. ASCL1, also known as MASH-1, is a basic helix-loop-helix (BHLH) transcription factor essential for the differentiation of neuroendocrine cells and neural tissues. It functions as a pioneer transcription factor that accesses closed chromatin to promote the transcription of neuronal genes.
  • Validated applications include western blotting, immunoprecipitation, and immunofluorescence (immunocytochemistry).
  • Species reactivity is confirmed for human, mouse, and rat.
  • Provided in a stable formulation with glycerol for storage at −20°C.
  • Recognizes endogenous levels of the 30 kDa ASCL1 protein.