Learn More
Aviva Systems Biology Corp ASCL2 Antibody - middle region (ARP32025_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32025_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ASCL2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ASCL2. Alternate names: ASH2, HASH2, MASH2, bHLHa45ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-54426