Learn More
Aviva Systems Biology Corp ASCL3 Antibody - C-terminal region (ARP39420_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39420_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ASCL3. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human ASCL3. Alternate names: HASH3, SGN1, bHLHa42ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-284-803