Learn More
Cayman Chemical AscochlorIn 2.5mg
SDP
Catalog No. 502983504
Encompass_Preferred
An isoprenoid antibiotic and antiviral that suppresses PMA-induced invasion in renal carcinoma cells (IC50 = ~10 µM) by inhibiting the expression of MMP-9; profoundly increases the expression of p53 by increasing protein stability in cancer cells and inhibits signaling through STAT3
Catalog No. 50-298-3504 Supplier Cayman Chemical Supplier No. 194372.5 MG
An isoprenoid antibiotic and antiviral that suppresses PMA-induced invasion in renal carcinoma cells (IC50 = ~10 µM) by inhibiting the expression of MMP-9; profoundly increases the expression of p53 by increasing protein stability in cancer cells and inhibits signaling through STAT3