Learn More
TARGETMOL CHEMICALS INC Ascomycin 500MG
SDP
Catalog No. 502877538
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Ascomycin (Immunomycin)(Immunomycin, FR-900520, FK520) is an ethyl analog of tacrolimus (FK506) with strong immunosuppressant properties. Purity 99.94%
Catalog No. 50-287-7538 Supplier TARGETMOL CHEMICALS INC Supplier No. T2481500MG
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Ascomycin (Immunomycin)(Immunomycin, FR-900520, FK520) is an ethyl analog of tacrolimus (FK506) with strong immunosuppressant properties. Purity 99.94%