Learn More
Abcam Ascomycin, Calcineurin/PP2B inhibitor, 1MG
Catalog No. 502706966
Encompass_Preferred
MW 792 Da, Purity >98%. Calcineurin/PP2B inhibitor (IC₅₀ = 49 nM). Forms a FK-506 binding protein 12 (FKBP12) - Ascomycin -Calcineurin complex. Immunosuppressant and antifungal activity in vitro and in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6966 Supplier Abcam Supplier No. AB1410161MG
MW 792 Da, Purity >98%. Calcineurin/PP2B inhibitor (IC₅₀ = 49 nM). Forms a FK-506 binding protein 12 (FKBP12) - Ascomycin -Calcineurin complex. Immunosuppressant and antifungal activity in vitro and in vivo.
The product is subject to the following: Abcam Restricted Use Statement