Learn More
AdipoGen Ascomycin high purity
SDP
Catalog No. 502562540
Encompass_Preferred
Chemical. CAS 104987-12-4. Formula C43H69NO12. MW 792. Isolated from Streptomyces sp. Macrolide antibiotic. Ethyl analog of FK506. Strong immunosuppressant. Binds to the FK-506-binding protein FKBP12. This complex inhibits calcineurin protein phosphatase 2B PP2B. Suppresses the production of T helper type 1 Th1 interferon and IL-2 and Th2 IL-4 and IL-10 cytokines in T lymphocytes and preferentially inhibits the activation of mast cells. Anti-inflammatory. Used for topical treatment of inflammatory skin diseases. Potently inhibits anti-IgE-induced histamine and cytokine release and reduces IgE-dependent p38 MAPK activation in human basophils. Inhibits basophil degranulation at the initial phase of allergic skin reactions. Antifungal. Antimalaria compound. Anticonvulsant antiepileptic.
Catalog No. 50-256-2540 Supplier AdipoGen Supplier No. AGCN20420M005
Chemical. CAS 104987-12-4. Formula C43H69NO12. MW 792. Isolated from Streptomyces sp. Macrolide antibiotic. Ethyl analog of FK506. Strong immunosuppressant. Binds to the FK-506-binding protein FKBP12. This complex inhibits calcineurin protein phosphatase 2B PP2B. Suppresses the production of T helper type 1 Th1 interferon and IL-2 and Th2 IL-4 and IL-10 cytokines in T lymphocytes and preferentially inhibits the activation of mast cells. Anti-inflammatory. Used for topical treatment of inflammatory skin diseases. Potently inhibits anti-IgE-induced histamine and cytokine release and reduces IgE-dependent p38 MAPK activation in human basophils. Inhibits basophil degranulation at the initial phase of allergic skin reactions. Antifungal. Antimalaria compound. Anticonvulsant antiepileptic.