Learn More
Cayman Chemical Ascorbate asy Buffer 25mL
SDP
Supplier:  Cayman Chemical 70042225 ML
Encompass_Preferred
A glutaminyl cyclase inhibitor (IC50 53 nM) reduces polyglutamine aggregation in HeLa cells expressing EGFP-HTT-Q74 at 12.5 and 25 UM reduces polyglutamine aggregation in primary mouse neurons expressing EGFP-Q80 at 50 UM
Catalog No. 50-289-6163