Learn More
Cayman Chemical Ascorbate asy KIt 96Wel
SDP
Catalog No. 502894163
Encompass_Preferred
Ascorbate (L-Ascorbic acid or Vitamin C) is a six-carbon lactone that is synthesized from glucose in the liver of most mammalian species, but not by humans who must obtain ascorbate through dietary consumption. In humans, ascorbate serves as an antioxidant and acts as an electron donor for various enzymes. Cayman’s Ascorbate Assay provides a reproducible, sensitive fluorescence-based tool for quantifying ascorbate from plasma, serum, urine, and fruit juices.
Catalog No. 50-289-4163 Supplier Cayman Chemical Supplier No. 70042096 WELL
Ascorbate (L-Ascorbic acid or Vitamin C) is a six-carbon lactone that is synthesized from glucose in the liver of most mammalian species, but not by humans who must obtain ascorbate through dietary consumption. In humans, ascorbate serves as an antioxidant and acts as an electron donor for various enzymes. Cayman’s Ascorbate Assay provides a reproducible, sensitive fluorescence-based tool for quantifying ascorbate from plasma, serum, urine, and fruit juices.