Learn More
Cayman Chemical Ascorbate DTPA 0.5mL
SDP
Catalog No. 502896922
Encompass_Preferred
A 1-opioid receptor antagonist (Ki 0.1 nM) selective for the 1-opioid receptor over the 2- - and K-opioid receptors (Kis 10.8 13.3 and 58.6 nM respectively) decreases proliferation of primary mouse splenic B cells and the production of IL-2 and IL-4 by primary mouse splenic T lymphocytes at 1 and 10 UM has antinociceptive activity in the tail-flick test in mice (ED50 46.4 pmol/animal) and decreases the antinociceptive activity of DPDPE at 6.3 pmol/animal decreases the number of capsaicin-induced coUGhs in mice at 0.3-3 mg/kg reduces the number of parasitized red blood cells in a mouse model of chloroquine-resistant P. chabaudi infection at 10 mg/kg
Catalog No. 50-289-6922 Supplier Cayman Chemical Supplier No. 7004230.5 ML
A 1-opioid receptor antagonist (Ki 0.1 nM) selective for the 1-opioid receptor over the 2- - and K-opioid receptors (Kis 10.8 13.3 and 58.6 nM respectively) decreases proliferation of primary mouse splenic B cells and the production of IL-2 and IL-4 by primary mouse splenic T lymphocytes at 1 and 10 UM has antinociceptive activity in the tail-flick test in mice (ED50 46.4 pmol/animal) and decreases the antinociceptive activity of DPDPE at 6.3 pmol/animal decreases the number of capsaicin-induced coUGhs in mice at 0.3-3 mg/kg reduces the number of parasitized red blood cells in a mouse model of chloroquine-resistant P. chabaudi infection at 10 mg/kg