Learn More
Medchemexpress LLC Ascorbate oxidase, Cucurbit sp. | 9029-44-1 | ≥100 U/mg solid | 1000 U
SDP
Catalog No. 5000400376
Encompass_Preferred
Ascorbate oxidase, Cucurbit sp., also known as vitamin C oxidase, is a REDOX enzyme involved in the regulation of the extracellular matrix. It catalyzes the reaction of ascorbic acid and oxygen to produce dehydroascorbic acid.
  • Solid appearance
  • White to off-white color
  • EC number: 1.10.3.3
  • Enzyme activity of ≥100 U/mg solid
  • Unit defined as the amount of enzyme that will oxidize 1.0 μmol ascorbic acid per min at 30°C, pH 5.6
Catalog No. 50-004-00376 Supplier Medchemexpress LLC Supplier No. HYP27421000U
Ascorbate oxidase, Cucurbit sp., also known as vitamin C oxidase, is a REDOX enzyme involved in the regulation of the extracellular matrix. It catalyzes the reaction of ascorbic acid and oxygen to produce dehydroascorbic acid.
  • Solid appearance
  • White to off-white color
  • EC number: 1.10.3.3
  • Enzyme activity of ≥100 U/mg solid
  • Unit defined as the amount of enzyme that will oxidize 1.0 μmol ascorbic acid per min at 30°C, pH 5.6