Learn More
Medchemexpress LLC Ascorbate oxidase, Cucurbit sp. | 9029-44-1 | ≥100 U/mg solid | 500 U
SDP
Catalog No. 5000400288
Encompass_Preferred
Ascorbate oxidase, Cucurbit sp., also known as vitamin C oxidase, is a REDOX enzyme involved in the regulation of the extracellular matrix. It catalyzes the reaction of ascorbic acid and oxygen to produce dehydroascorbic acid.
  • REDOX enzyme involved in the regulation of extracellular matrix.
  • Catalyzes the reaction of ascorbic acid and oxygen to produce dehydroascorbic acid.
  • For research use only.
  • Available in 100 U, 500 U, and 1000 U quantities.
  • Purity (Enzyme Activity) is ≥100 U/mg solid.
Catalog No. 50-004-00288 Supplier Medchemexpress LLC Supplier No. HYP2742500U
Ascorbate oxidase, Cucurbit sp., also known as vitamin C oxidase, is a REDOX enzyme involved in the regulation of the extracellular matrix. It catalyzes the reaction of ascorbic acid and oxygen to produce dehydroascorbic acid.
  • REDOX enzyme involved in the regulation of extracellular matrix.
  • Catalyzes the reaction of ascorbic acid and oxygen to produce dehydroascorbic acid.
  • For research use only.
  • Available in 100 U, 500 U, and 1000 U quantities.
  • Purity (Enzyme Activity) is ≥100 U/mg solid.