Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ascorbate oxidase, Cucurbit sp. | 9029-44-1 | 100 U
SDP

Catalog No. 5000400290
Encompass_Preferred

Ascorbate oxidase, Cucurbit sp. is a REDOX enzyme involved in the regulation of extracellular matrix. It catalyzes the reaction of ascorbic acid and oxygen to produce dehydroascorbic acid. Also known as vitamin C oxidase, this enzyme is provided as a white to off-white solid.

  • Catalyzes the reaction of ascorbic acid and oxygen to produce dehydroascorbic acid
  • Functions as a REDOX enzyme
  • Involved in the regulation of extracellular matrix
  • EC number: 1.10.3.3
  • Enzyme activity: ≥100 U/mg solid
  • Unit defined as the amount of enzyme that oxidizes 1.0 μmol ascorbic acid per minute at 30°C, pH 5.6
  • For research use only

Catalog No. 50-004-00290 Supplier Medchemexpress LLC Supplier No. HYP2742100U
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ascorbate oxidase, Cucurbit sp. is a REDOX enzyme involved in the regulation of extracellular matrix. It catalyzes the reaction of ascorbic acid and oxygen to produce dehydroascorbic acid. Also known as vitamin C oxidase, this enzyme is provided as a white to off-white solid.

  • Catalyzes the reaction of ascorbic acid and oxygen to produce dehydroascorbic acid
  • Functions as a REDOX enzyme
  • Involved in the regulation of extracellular matrix
  • EC number: 1.10.3.3
  • Enzyme activity: ≥100 U/mg solid
  • Unit defined as the amount of enzyme that oxidizes 1.0 μmol ascorbic acid per minute at 30°C, pH 5.6
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.