Learn More
Sigma Aldrich Fine Chemicals Biosciences Ascorbate Oxidase from Cuc
Catalog No. 502903325
Encompass
Ascorbate oxidase is a homodimeric enzyme which comprises 552 amino acid residues per subunit (zucchini). It corresponds to a molecular mass of 70kDa per subunit. This enzyme is mainly found in plants fungi and eubacteria.
Catalog No. 50-290-3325 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. A01571KU
Ascorbate oxidase is a homodimeric enzyme which comprises 552 amino acid residues per subunit (zucchini). It corresponds to a molecular mass of 70kDa per subunit. This enzyme is mainly found in plants fungi and eubacteria.