Learn More
MOLECULAR DEPOT LLC Ascorbic Acid - 2 /1 mL
Catalog No. 5000013495
Encompass_Preferred
Ascorbic Acid for Interference Testing consists of a highly concentrated solution of L-ascorbic acid (176 mg/mL - 1 0 M) provided with the corresponding diluent (diluent A) This solution can be spiked into specimens to test ascorbic acid interference/tolerance in assays Instructions and sample calculations are provided For in vitro research use only
Catalog No. 50-000-13495 Supplier MOLECULAR DEPOT LLC Supplier No. K2010001A
Ascorbic Acid for Interference Testing consists of a highly concentrated solution of L-ascorbic acid (176 mg/mL - 1 0 M) provided with the corresponding diluent (diluent A) This solution can be spiked into specimens to test ascorbic acid interference/tolerance in assays Instructions and sample calculations are provided For in vitro research use only