Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ascorbic Acid, 2% (w/v), Ricca Chemical
SDP

Catalog No. R08082001C Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R08082001C 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R08082001C Supplier Ricca Chemical Company Supplier No. R08082001C
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7732-18-5 , 50-81-7
Synonym dihydrogen oxide, dihydrogen monoxide

Specifications

Boiling Point Approximately 100°C
Odor Odorless
Packaging Amber Glass Bottle
Quantity 1 L
Solubility Information Solubility in Water: Infinite
Percent Purity 2% w/v
Grade Laboratory
Physical Form Liquid
Recommended Storage General
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.