Learn More
Lifeline Cell Technology Ascorbic Acid LifeFactor - 0.5 mL
SDP
Catalog No. NC3454733
Encompass_Preferred
May be used with:FibroLife® basal medium and human fibroblastsVascuLife® basal medium and human smooth muscle cellsVascuLife® basal medium and human endothelial cellsDermaLife basal medium and human melanocytes (neonatal, adult)
Catalog No. NC3454733 Supplier Lifeline Cell Technology Supplier No. LS1005
May be used with:FibroLife® basal medium and human fibroblastsVascuLife® basal medium and human smooth muscle cellsVascuLife® basal medium and human endothelial cellsDermaLife basal medium and human melanocytes (neonatal, adult)