Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ascorbic Acid, Pharmaceutical Secondary Standard, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111014384 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4384 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 11-101-4384 Supplier MilliporeSigma Supelco Supplier No. PHR100810G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Ascorbic acid is a powerful antioxidant and is widely used in pharmaceutical formulations. It has proven to have a beneficial effect on age-related diseases such as cancer, atherosclerosis, ocular, neurodegenerative diseases, etc. It is mainly involved in the formation and maintenance of collagen. The prolonged deficiency of ascorbic acid in diet may result in scurvy.
TRUSTED_SUSTAINABILITY

Specifications

CAS 50-81-7
Linear Formula C6H8O6
MDL Number MFCD00064328
Melting Point 190°C to 194°C (decomposition)
Synonym L-Ascorbic acid; L-Threoascorbic acid; Antiscorbutic factor; Vitamin C
Quantity 10 g
Formula Weight 176.12
Grade Certified Reference Material
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.