Learn More
Hach Company Ascorbic Acid Reagent Powder for Phosphorus, 48 g
Supplier:  Hach Company 2651255
Encompass
For determination of reactive phosphorus (phosphate) by the Rapid Liquid Ascorbic Acid Method. Hach Method 10055. Used with Ascorbic Acid Reagent Dilution Solution (2599949) and Molybdate Reagent Solution (2599849). 48 g bottle.
Catalog No. 50-199-6436