Learn More
Hach Company Ascorbic Acid Powder Pillows, pk/100
Supplier:  Hach Company 1457799
Encompass
Reagent for aluminum determination by the aluminon method (Method 8012), manganese determination by the PAN method (Method 8149) and total chelant determination by titration (Method 8350). Pack of 100 powder pillows.
Catalog No. 50-199-5086