Learn More
Ascorbic Acid Testing Solution, Reagents
Specifications
Specifications
| Chemical Name or Material | Ascorbic Acid Testing Solution |
| Color | Colorless to Light Yellow |
| CAS | 7732-18-5, 7647-01-0, 7647-01-0 |
| Boiling Point | 100°C |
| Grade | Reagent |
| Specific Gravity | 1.0039 |
| pH | <1 |
| Quantity | 1 L |
| Sterility | Non-sterile |
| Physical Form | Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.