Learn More
Hach Company Ascorbic Acid Titrant, 500 mL
Catalog No. 501995770 Shop All Hach Company Products
Encompass
For determination of ascorbic acid in juice based beverages, by drop-count titration. 500 mL.bottle
Catalog No. 50-199-5770 Supplier Hach Company Supplier No. 2308249
For determination of ascorbic acid in juice based beverages, by drop-count titration. 500 mL.bottle