Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medisca Ascorbic Acid, USP/EP (Dietary Supplement Grade)
SDP

Catalog No. 14185616 Shop All Medisca Products
Change view
Click to view available options
Quantity:
500 g
1 kg
25 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-185-616 500 g
14-185-547 1 kg
14-185-549 25 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14-185-616 Supplier Medisca Supplier No. 063408
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ascorbic Acid, USP/EP (Dietary Supplement Grade) ranges from white to slightly yellow in colour, and comes either as crystals or powder in texture.

Ascorbic Acid USP/EP ranges from white to slightly yellow in colour, and comes either as crystals or powder in texture. The product is light-sensitive and on exposure to light will gradually darken. In the dry state, Ascorbic Acid is reasonably stable in air but in solution it rapidly oxidizes.

Dietary Supplement Grade refers to a nutritional supplement that is manufactured by an FDA-registered dietary supplement manufacturer or manufacturer with other certifications and registrations, based on applicable laws and regulations. Qualified manufacturers follow their applicable standards of practice, which are less stringent than those for FDA-registered drug manufacturers. The customer is responsible for ensuring suitability of the product for the intended use and acknowledges that further testing may be required.

Alternative name(s): Vitamin C

Specifications

Chemical Name or Material Ascorbic Acid, USP/EP (Dietary Supplement Grade)
Packaging Jar, 600mL, White, HDPE, Round, 89-400, TE Pano Neck
CAS 50-81-7
Type Drug Standard
Molecular Weight (g/mol) 176.12
Quantity 500 g
Percent Purity 99 to 100.5 %
Grade EP, USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.