Learn More
Medchemexpress LLC Ascorbyl palmitate | 137-66-6 | 99.62% | 500 MG
SDP
Catalog No. 5000409153
Encompass_Preferred
Ascorbyl palmitate is an orally active ester formed from ascorbic acid and palmitic acid. It functions as an antioxidant and food additive, more efficient than other common antioxidants in preventing fat and oil oxidation. It also plays a role in mitigating the inhibition of collagen synthesis and has been shown to induce apoptosis in HUVECs and ameliorate inflammatory diseases.
  • Functions as an antioxidant and food additive
  • More efficient in preventing fat and oil oxidation
  • Mitigates inhibition of collagen synthesis
  • Induces apoptosis in human umbilical vein endothelial cells (HUVECs)
  • Ameliorates inflammatory diseases by inhibiting the NLRP3 inflammasome
Catalog No. 50-004-09153 Supplier Medchemexpress LLC Supplier No. HYB0987500MG
Ascorbyl palmitate is an orally active ester formed from ascorbic acid and palmitic acid. It functions as an antioxidant and food additive, more efficient than other common antioxidants in preventing fat and oil oxidation. It also plays a role in mitigating the inhibition of collagen synthesis and has been shown to induce apoptosis in HUVECs and ameliorate inflammatory diseases.
  • Functions as an antioxidant and food additive
  • More efficient in preventing fat and oil oxidation
  • Mitigates inhibition of collagen synthesis
  • Induces apoptosis in human umbilical vein endothelial cells (HUVECs)
  • Ameliorates inflammatory diseases by inhibiting the NLRP3 inflammasome