Learn More
Cayman Chemical Ascorbyl Tetraisopalmitate, Purity: ≥95%, CAS: 183476-82-6, 5 g
SDP
Catalog No. 502912929
Encompass_Preferred
A vitamin C derivative; protects against UVB-, hydrogen peroxide-, or tert-butyl hydroperoxide-induced decreases in cell viability in HaCaT keratinocytes at 100 µM; decreases IL-1α and PGE2 production in unstimulated and UVB-stimulated HaCaT keratinocytes at 20 µM; topical application increases viable epidermis thickness, stratum corneum water content, and skin smoothness and decreases skin roughness in hairless mice
Catalog No. 50-291-2929 Supplier Cayman Chemical Supplier No. 362755 G
A vitamin C derivative; protects against UVB-, hydrogen peroxide-, or tert-butyl hydroperoxide-induced decreases in cell viability in HaCaT keratinocytes at 100 µM; decreases IL-1α and PGE2 production in unstimulated and UVB-stimulated HaCaT keratinocytes at 20 µM; topical application increases viable epidermis thickness, stratum corneum water content, and skin smoothness and decreases skin roughness in hairless mice