Learn More
Cell Signaling Technology Matched Antibody Pair, AsCpf1/Cas12a, Strain BV3L6, 1 Kit
SDP
Catalog No. 503132466
Encompass
The AsCpf1/Cas12a (Strain BV3L6) Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms that require specialized or custom antibody labeling. This kit includes capture and detection antibodies that bind to non-overlapping epitopes and are supplied in a buffer free of BSA and azide, facilitating the conjugation of labels such as fluorophores, lanthanides, biotin, and beads for use on platforms including MSD, Quanterix Simoa, Luminex, and Alpha Technology.
  • Includes one capture antibody and one detection antibody, each at 100 μg.
  • Optimized for versatility with platforms such as MSD, Quanterix Simoa, Alpha Technology, and Luminex.
  • Supplied in 1X PBS (10 mM Na2HPO4, 3 mM KCl, 2 mM KH2PO4, and 140 mM NaCl, pH 7.8).
  • BSA and azide free formulation.
  • Validated for ELISA and ELISA-like assays.
Catalog No. 50-313-2466 Supplier Cell Signaling Technology Supplier No. 94099S
The AsCpf1/Cas12a (Strain BV3L6) Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms that require specialized or custom antibody labeling. This kit includes capture and detection antibodies that bind to non-overlapping epitopes and are supplied in a buffer free of BSA and azide, facilitating the conjugation of labels such as fluorophores, lanthanides, biotin, and beads for use on platforms including MSD, Quanterix Simoa, Luminex, and Alpha Technology.
  • Includes one capture antibody and one detection antibody, each at 100 μg.
  • Optimized for versatility with platforms such as MSD, Quanterix Simoa, Alpha Technology, and Luminex.
  • Supplied in 1X PBS (10 mM Na2HPO4, 3 mM KCl, 2 mM KH2PO4, and 140 mM NaCl, pH 7.8).
  • BSA and azide free formulation.
  • Validated for ELISA and ELISA-like assays.