Learn More
Medchemexpress LLC HY-N8393 1mg , Ascr#18 CAS:1355681-10-5 Purity:>98%
SDP
Catalog No. 502261559
Encompass_Preferred
HY-N8393 1mg Ascr#18 CAS: 1355681-10-5 Ascr#18, an ascaroside, is a hormone of nematodes. Ascr#18 is expressed during nematode development. Ascr#18 increases resistance in Arabidopsis, tomato, potato and barley to viral, bacterial, oomycete, fungal and nematode infections. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-226-1559 Supplier Medchemexpress LLC Supplier No. HYN83931MG
HY-N8393 1mg Ascr#18 CAS: 1355681-10-5 Ascr#18, an ascaroside, is a hormone of nematodes. Ascr#18 is expressed during nematode development. Ascr#18 increases resistance in Arabidopsis, tomato, potato and barley to viral, bacterial, oomycete, fungal and nematode infections. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.